Product Details

SNP ID
rs1826607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:114097928 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACAAATAATTGAAGAATCCTAGA[A/G]TTGAAAGGAAACTTTGAGATCATGA
Phenotype
MIM: 605317
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOXP2 PubMed Links

Gene Details

Gene
FOXP2
Gene Name
forkhead box P2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172766.2 Intron NP_001166237.1
NM_001172767.2 Intron NP_001166238.1
NM_014491.3 Intron NP_055306.1
NM_148898.3 Intron NP_683696.2
NM_148899.3 Intron NP_683697.2
NM_148900.3 Intron NP_683698.2
XM_017012801.1 Intron XP_016868290.1

View Full Product Details