Product Details

SNP ID
rs1868902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:172735551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGCCCTCTCCCCACTCTGCCT[C/T]GGCGCCTGCCCTCCCGCCCCGCCTC
Phenotype
MIM: 606058
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAPGEF4 PubMed Links

Gene Details

Gene
RAPGEF4
Gene Name
Rap guanine nucleotide exchange factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100397.1 Intron NP_001093867.1
NM_001282899.1 Intron NP_001269828.1
NM_001282900.1 Intron NP_001269829.1
NM_001282901.1 Intron NP_001269830.1
NM_007023.3 Intron NP_008954.2
XM_005246246.2 Intron XP_005246303.1
XM_006712205.3 Intron XP_006712268.1
XM_011510505.2 Intron XP_011508807.1
XM_017003196.1 Intron XP_016858685.1
XM_017003197.1 Intron XP_016858686.1
Gene
RAPGEF4-AS1
Gene Name
RAPGEF4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details