Product Details

SNP ID
rs1880952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73666305 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGACAGTGTCCCCACAGGGCCTA[A/G]CCCAGTGTTCAGCGCAGAGCGGCAT
Phenotype
MIM: 610039
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS37D PubMed Links

Gene Details

Gene
VPS37D
Gene Name
VPS37D, ESCRT-I subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077621.1 Intron NP_001071089.1
XM_017011779.1 Intron XP_016867268.1

View Full Product Details