Product Details

SNP ID
rs1865679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:20670258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAAACCAAAGCTCGGTTCGGATA[G/T]CCCGGGCGAAGACGTCCGCTGCTCT
Phenotype
MIM: 604651
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GDF7 PubMed Links

Gene Details

Gene
GDF7
Gene Name
growth differentiation factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182828.3 Intron NP_878248.2

View Full Product Details