Product Details

SNP ID
rs1318008
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:12066308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAGGTTCGGGAAGCTCGTTATTT[A/T]AAAAAAAAAATCCCAAATTAGGGCT
Phenotype
MIM: 153243
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TNFRSF8 PubMed Links

Gene Details

Gene
TNFRSF8
Gene Name
TNF receptor superfamily member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243.4 Intron NP_001234.3
NM_001281430.2 Intron NP_001268359.2
XM_011542441.2 Intron XP_011540743.1
XM_011542443.2 Intron XP_011540745.1

View Full Product Details