Product Details

SNP ID
rs2249473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:10467215 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTCTCTATAAATTTTATAATTTC[A/T]GTGTTCACACACAGTTATATCATGC
Phenotype
MIM: 602538
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CELF2 PubMed Links

Gene Details

Gene
CELF2
Gene Name
CUGBP, Elav-like family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025076.2 Intron NP_001020247.1
NM_001025077.2 Intron NP_001020248.1
NM_001083591.1 Intron NP_001077060.1
NM_001326317.1 Intron NP_001313246.1
NM_001326318.1 Intron NP_001313247.1
NM_001326319.1 Intron NP_001313248.1
NM_001326320.1 Intron NP_001313249.1
NM_001326321.1 Intron NP_001313250.1
NM_001326323.1 Intron NP_001313252.1
NM_001326324.1 Intron NP_001313253.1
NM_001326325.1 Intron NP_001313254.1
NM_001326326.1 Intron NP_001313255.1
NM_001326327.1 Intron NP_001313256.1
NM_001326328.1 Intron NP_001313257.1
NM_001326329.1 Intron NP_001313258.1
NM_001326330.1 Intron NP_001313259.1
NM_001326331.1 Intron NP_001313260.1
NM_001326332.1 Intron NP_001313261.1
NM_001326333.1 Intron NP_001313262.1
NM_001326334.1 Intron NP_001313263.1
NM_001326335.1 Intron NP_001313264.1
NM_001326336.1 Intron NP_001313265.1
NM_001326337.1 Intron NP_001313266.1
NM_001326338.1 Intron NP_001313267.1
NM_001326339.1 Intron NP_001313268.1
NM_001326340.1 Intron NP_001313269.1
NM_001326341.1 Intron NP_001313270.1
NM_001326342.1 Intron NP_001313271.1
NM_001326343.1 Intron NP_001313272.1
NM_001326344.1 Intron NP_001313273.1
NM_001326345.1 Intron NP_001313274.1
NM_001326346.1 Intron NP_001313275.1
NM_001326347.1 Intron NP_001313276.1
NM_001326348.1 Intron NP_001313277.1
NM_001326349.1 Intron NP_001313278.1
NM_006561.3 Intron NP_006552.3
XM_005252354.4 Intron XP_005252411.1
XM_005252357.4 Intron XP_005252414.1
XM_005252358.3 Intron XP_005252415.1
XM_006717373.2 Intron XP_006717436.1
XM_011519294.2 Intron XP_011517596.1
XM_011519297.1 Intron XP_011517599.1
XM_017015542.1 Intron XP_016871031.1
XM_017015543.1 Intron XP_016871032.1
XM_017015544.1 Intron XP_016871033.1
XM_017015545.1 Intron XP_016871034.1
XM_017015546.1 Intron XP_016871035.1
XM_017015547.1 Intron XP_016871036.1
XM_017015548.1 Intron XP_016871037.1
XM_017015549.1 Intron XP_016871038.1
XM_017015550.1 Intron XP_016871039.1
XM_017015551.1 Intron XP_016871040.1
XM_017015552.1 Intron XP_016871041.1
XM_017015553.1 Intron XP_016871042.1
XM_017015554.1 Intron XP_016871043.1
XM_017015555.1 Intron XP_016871044.1
XM_017015556.1 Intron XP_016871045.1
XM_017015557.1 Intron XP_016871046.1
XM_017015558.1 Intron XP_016871047.1
XM_017015559.1 Intron XP_016871048.1
XM_017015560.1 Intron XP_016871049.1
XM_017015561.1 Intron XP_016871050.1
XM_017015562.1 Intron XP_016871051.1
XM_017015563.1 Intron XP_016871052.1
XM_017015564.1 Intron XP_016871053.1
XM_017015565.1 Intron XP_016871054.1
XM_017015566.1 Intron XP_016871055.1
XM_017015567.1 Intron XP_016871056.1
XM_017015568.1 Intron XP_016871057.1
XM_017015569.1 Intron XP_016871058.1
XM_017015570.1 Intron XP_016871059.1
XM_017015571.1 Intron XP_016871060.1
XM_017015572.1 Intron XP_016871061.1
Gene
LOC101928322
Gene Name
uncharacterized LOC101928322
There are no transcripts associated with this gene.

View Full Product Details