Product Details

SNP ID
rs5547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:40113814 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATAAGGCCTTTCTCCTTGGCCTT[C/G]CTGAATTAATGGGTGATTTTTAGAG
Phenotype
MIM: 182305
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC8A1 PubMed Links

Gene Details

Gene
SLC8A1
Gene Name
solute carrier family 8 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112800.1 4570 UTR 3 NP_001106271.1
NM_001112801.1 4570 UTR 3 NP_001106272.1
NM_001112802.1 4570 UTR 3 NP_001106273.1
NM_001252624.1 4570 UTR 3 NP_001239553.1
NM_021097.2 4570 UTR 3 NP_066920.1
XM_005264514.3 4570 UTR 3 XP_005264571.1
XM_006712081.2 4570 UTR 3 XP_006712144.1
XM_006712082.3 4570 UTR 3 XP_006712145.1
XM_006712083.3 4570 UTR 3 XP_006712146.1
XM_006712084.3 4570 UTR 3 XP_006712147.1
XM_006712085.3 4570 UTR 3 XP_006712148.1
XM_011533050.1 4570 UTR 3 XP_011531352.1
XM_011533054.2 4570 UTR 3 XP_011531356.1
XM_011533055.2 4570 UTR 3 XP_011531357.1
XM_011533056.1 4570 UTR 3 XP_011531358.1
XM_011533057.2 4570 UTR 3 XP_011531359.2
XM_011533058.2 4570 Intron XP_011531360.1
XM_017004745.1 4570 UTR 3 XP_016860234.1
XM_017004746.1 4570 UTR 3 XP_016860235.1
XM_017004747.1 4570 UTR 3 XP_016860236.1
XM_017004748.1 4570 UTR 3 XP_016860237.1
XM_017004749.1 4570 UTR 3 XP_016860238.1
XM_017004750.1 4570 UTR 3 XP_016860239.1
XM_017004751.1 4570 UTR 3 XP_016860240.1
XM_017004752.1 4570 UTR 3 XP_016860241.1
XM_017004753.1 4570 UTR 3 XP_016860242.1
XM_017004754.1 4570 UTR 3 XP_016860243.1
XM_017004755.1 4570 UTR 3 XP_016860244.1
XM_017004756.1 4570 UTR 3 XP_016860245.1
XM_017004757.1 4570 UTR 3 XP_016860246.1
XM_017004758.1 4570 UTR 3 XP_016860247.1
XM_017004759.1 4570 UTR 3 XP_016860248.1
XM_017004760.1 4570 UTR 3 XP_016860249.1
XM_017004761.1 4570 UTR 3 XP_016860250.1
XM_017004762.1 4570 UTR 3 XP_016860251.1
XM_017004763.1 4570 UTR 3 XP_016860252.1
XM_017004764.1 4570 Intron XP_016860253.1
Gene
SLC8A1-AS1
Gene Name
SLC8A1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details