Product Details

SNP ID
rs2033173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:25741377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAACAGTACCAGTAACTAGTAAC[C/T]AGTACAGTAGAAGCCAAGAACCAAG
Phenotype
MIM: 612991
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASXL2 PubMed Links

Gene Details

Gene
ASXL2
Gene Name
additional sex combs like 2, transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018263.4 4588 UTR 3 NP_060733.4
XM_006712039.3 4588 UTR 3 XP_006712102.1
XM_006712040.2 4588 UTR 3 XP_006712103.1
XM_011532950.2 4588 UTR 3 XP_011531252.1
XM_011532951.2 4588 UTR 3 XP_011531253.1
XM_017004429.1 4588 UTR 3 XP_016859918.1
XM_017004430.1 4588 UTR 3 XP_016859919.1

View Full Product Details