Product Details

SNP ID
rs2359425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:76862211 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAAAAGTAGAGTAGCCTAATGAA[A/C]TCCCATGTACCCAGCTGAAGAAGGA
Phenotype
MIM: 610103
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
S100Z PubMed Links

Gene Details

Gene
S100Z
Gene Name
S100 calcium binding protein Z
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130772.3 Intron NP_570128.2
XM_011543239.2 Intron XP_011541541.1
XM_011543240.2 Intron XP_011541542.1
XM_011543241.2 Intron XP_011541543.1
XM_011543244.2 Intron XP_011541546.1
XM_011543245.2 Intron XP_011541547.1
XM_017009171.1 Intron XP_016864660.1
XM_017009172.1 Intron XP_016864661.1
XM_017009173.1 Intron XP_016864662.1

View Full Product Details