Product Details

SNP ID
rs1952424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:99690434 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTCTCCGCCTTTCCCTGGGAGAA[C/T]GTGATTTTGTTGTCACCCATTGGGG
Phenotype
MIM: 604087
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP46A1 PubMed Links

Gene Details

Gene
CYP46A1
Gene Name
cytochrome P450 family 46 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006668.1 Intron NP_006659.1
XM_005267274.4 Intron XP_005267331.1
XM_011536364.1 Intron XP_011534666.1
XM_011536365.1 Intron XP_011534667.1
XM_017020933.1 Intron XP_016876422.1
Gene
HHIPL1
Gene Name
HHIP like 1
There are no transcripts associated with this gene.

View Full Product Details