Product Details

SNP ID
rs2617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103733971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACACACGACTCATACATGAAAAA[A/T]AGAGCCTAAGGGCCTGTATTTTAAT
Phenotype
MIM: 606455 MIM: 613504
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PPP1R13B PubMed Links

Gene Details

Gene
PPP1R13B
Gene Name
protein phosphatase 1 regulatory subunit 13B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015316.2 5248 UTR 3 NP_056131.2
XM_005267487.4 5248 UTR 3 XP_005267544.1
XM_011536593.2 5248 UTR 3 XP_011534895.1
XM_017021116.1 5248 UTR 3 XP_016876605.1
XM_017021117.1 5248 UTR 3 XP_016876606.1
XM_017021118.1 5248 UTR 3 XP_016876607.1
XM_017021119.1 5248 UTR 3 XP_016876608.1
Gene
ZFYVE21
Gene Name
zinc finger FYVE-type containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198953.1 5248 Intron NP_001185882.1
NM_024071.3 5248 Intron NP_076976.1

View Full Product Details