Product Details

SNP ID
rs2053588
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:96379755 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACCTGTGAGGATTCCTTCCCTTCA[A/G]GTACTGGATTCTTGATCTTTCTGCA
Phenotype
MIM: 616605
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSKIP PubMed Links

Gene Details

Gene
GSKIP
Gene Name
GSK3B interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271904.1 185 UTR 5 NP_001258833.1
NM_001271905.1 185 UTR 5 NP_001258834.1
NM_001271906.1 185 Intron NP_001258835.1
NM_016472.4 185 UTR 5 NP_057556.2
XM_017021365.1 185 UTR 5 XP_016876854.1

View Full Product Details