Product Details

SNP ID
rs1885419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:95652552 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGTTTTCCTTTAGAGTTGGGCAG[C/G]GCCTGGCTGGACTTCTTTCTCCACC
Phenotype
MIM: 604412
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TCL6 PubMed Links

Gene Details

Gene
TCL6
Gene Name
T-cell leukemia/lymphoma 6 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details