Product Details

SNP ID
rs10197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:91869577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAGCATGTGTCTGAAGGCCTTTC[A/G]AAGGAATTCTACGACATATTTTTTA
Phenotype
MIM: 604580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBLN5 PubMed Links

Gene Details

Gene
FBLN5
Gene Name
fibulin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006329.3 2557 UTR 3 NP_006320.2
XM_005267267.3 2557 UTR 3 XP_005267324.1
XM_011536356.1 2557 UTR 3 XP_011534658.1
XM_011536357.1 2557 UTR 3 XP_011534659.1
XM_011536358.1 2557 UTR 3 XP_011534660.1
XM_017020929.1 2557 UTR 3 XP_016876418.1
Gene
TC2N
Gene Name
tandem C2 domains, nuclear
There are no transcripts associated with this gene.

View Full Product Details