Product Details

SNP ID
rs35930119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:48306569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAGATCTTCCTCCTTACAGCCA[C/T]GGCCTATGACCGCTATGTGGCCATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4S1 PubMed Links

Gene Details

Gene
OR4S1
Gene Name
olfactory receptor family 4 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004725.1 347 Missense Mutation ACG,ATG T116M NP_001004725.1

View Full Product Details