Product Details

SNP ID
rs11166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68207005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTAACACCACTTGGGGCTGTGAC[C/T]GTATTTACTTCATTCTTGAATCCCG
Phenotype
MIM: 606725
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALML4 PubMed Links

Gene Details

Gene
CALML4
Gene Name
calmodulin like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031733.2 2229 Intron NP_001026903.2
NM_001286694.1 2229 Intron NP_001273623.1
NM_001286695.1 2229 Intron NP_001273624.1
NM_033429.2 2229 Intron NP_219501.2
Gene
CLN6
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017882.2 2229 UTR 3 NP_060352.1

View Full Product Details