Product Details

SNP ID
rs2014
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:16272336 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAAACAAAGCAGTTTTACTGTACA[A/C]AGAAGTGCAATGCTACATTAAGTCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIF16B PubMed Links

Gene Details

Gene
KIF16B
Gene Name
kinesin family member 16B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199865.1 4902 UTR 3 NP_001186794.1
NM_001199866.1 4902 Intron NP_001186795.1
NM_024704.4 4902 UTR 3 NP_078980.3
XM_005260750.3 4902 Intron XP_005260807.1
XM_005260751.4 4902 Intron XP_005260808.1
XM_005260752.3 4902 Intron XP_005260809.1
XM_005260753.3 4902 UTR 3 XP_005260810.1
XM_005260754.3 4902 UTR 3 XP_005260811.1
XM_005260755.3 4902 UTR 3 XP_005260812.1
XM_006723588.3 4902 UTR 3 XP_006723651.1
XM_017027926.1 4902 UTR 3 XP_016883415.1

View Full Product Details