Product Details

SNP ID
rs9726184
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:27736511 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGATACTAGTAAAACTCTTAGAA[C/T]GGAATCTGGTACATGAAAAATACTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM76A PubMed Links

Gene Details

Gene
FAM76A
Gene Name
family with sequence similarity 76 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143912.1 Intron NP_001137384.1
NM_001143913.1 Intron NP_001137385.1
NM_001143914.1 Intron NP_001137386.1
NM_001143915.1 Intron NP_001137387.1
NM_152660.2 Intron NP_689873.1
XM_017000543.1 Intron XP_016856032.1

View Full Product Details