Product Details

SNP ID
rs2052759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50183225 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGAGGTGAAGCCTCATGCCCAGC[C/T]ATCTATCTCCAAATTCCAAGCTCTT
Phenotype
MIM: 609123 MIM: 603247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
There are no transcripts associated with this gene.

Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159629.1 Intron NP_001153101.1
NM_003645.3 Intron NP_003636.2

View Full Product Details