Product Details

SNP ID
rs2024953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:9071342 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATATTTGGCAATTTCAGTAAACAT[G/T]GTATTTTCCTAAGATACCACTAACT
Phenotype
MIM: 600810
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLCB4 PubMed Links

Gene Details

Gene
PLCB4
Gene Name
phospholipase C beta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000933.3 Intron NP_000924.3
NM_001172646.1 Intron NP_001166117.1
NM_182797.2 Intron NP_877949.2
XM_005260724.2 Intron XP_005260781.1
XM_005260728.2 Intron XP_005260785.1
XM_006723568.3 Intron XP_006723631.1
XM_006723569.2 Intron XP_006723632.1
XM_011529254.2 Intron XP_011527556.1
XM_017027880.1 Intron XP_016883369.1
XM_017027881.1 Intron XP_016883370.1
XM_017027882.1 Intron XP_016883371.1
XM_017027883.1 Intron XP_016883372.1
XM_017027884.1 Intron XP_016883373.1
XM_017027885.1 Intron XP_016883374.1
XM_017027886.1 Intron XP_016883375.1

View Full Product Details