Product Details

SNP ID
rs10915578
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:4663857 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAGACTGTTTGCCGTGTCTGCAT[A/G]GGACATGGGATGAAAGAAGGGTGAC
Phenotype
MIM: 610972
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AJAP1 PubMed Links

Gene Details

Gene
AJAP1
Gene Name
adherens junctions associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042478.1 Intron NP_001035943.1
NM_018836.3 Intron NP_061324.1
XM_011541786.2 Intron XP_011540088.1
XM_011541787.2 Intron XP_011540089.1

View Full Product Details