Product Details
- SNP ID
-
rs14547
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:21595642 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCAGCCTGCGCGCCGCCTCGCCC[G/T]CCCCGTCCAGCGCGCCCTCGCACGC
- Phenotype
-
MIM: 184430
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SOX4
PubMed Links
Gene Details
- Gene
- SOX4
- Gene Name
- SRY-box 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003107.2 |
1902 |
Missense Mutation |
GCC,TCC |
A370S |
NP_003098.1 |
View Full Product Details