Product Details

SNP ID
rs739706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:47088883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAATATTTATTAGAACTACTGGA[C/T]GCCTAAATTCCTTTTGTCTCAATTT
Phenotype
MIM: 615690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMIGO2 PubMed Links

Gene Details

Gene
AMIGO2
Gene Name
adhesion molecule with Ig like domain 2
There are no transcripts associated with this gene.

Gene
PCED1B
Gene Name
PC-esterase domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281429.1 Intron NP_001268358.1
NM_138371.2 Intron NP_612380.1
XM_005269224.4 Intron XP_005269281.1
XM_011538978.2 Intron XP_011537280.1
XM_017020207.1 Intron XP_016875696.1
XM_017020208.1 Intron XP_016875697.1
XM_017020209.1 Intron XP_016875698.1
XM_017020210.1 Intron XP_016875699.1
XM_017020211.1 Intron XP_016875700.1
XM_017020212.1 Intron XP_016875701.1
XM_017020213.1 Intron XP_016875702.1
XM_017020214.1 Intron XP_016875703.1
XM_017020215.1 Intron XP_016875704.1
XM_017020216.1 Intron XP_016875705.1

View Full Product Details