Product Details

SNP ID
rs1886198
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:72783837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAATGATACATTTATATTTCTTG[A/T]CAAGTACAGCCTTTAAATAATTTTG
Phenotype
MIM: 607533 MIM: 607532
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
DIS3 PubMed Links

Gene Details

Gene
DIS3
Gene Name
DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease
There are no transcripts associated with this gene.

Gene
PIBF1
Gene Name
progesterone immunomodulatory binding factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006346.2 Intron NP_006337.2
XM_011534881.2 Intron XP_011533183.1
XM_011534882.2 Intron XP_011533184.1
XM_011534884.2 Intron XP_011533186.1
XM_011534885.2 Intron XP_011533187.1
XM_011534886.2 Intron XP_011533188.1
XM_017020350.1 Intron XP_016875839.1
XM_017020351.1 Intron XP_016875840.1
XM_017020352.1 Intron XP_016875841.1
XM_017020353.1 Intron XP_016875842.1
XM_017020354.1 Intron XP_016875843.1

View Full Product Details