Product Details
- SNP ID
-
rs35623356
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:12624515 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ATTTCTTGCTCATAATCACTGTTCC[A/G]AGATTTGGAATCTGAAAAGGAAACA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF791
PubMed Links
Gene Details
- Gene
- ZNF791
- Gene Name
- zinc finger protein 791
View Full Product Details