Product Details

SNP ID
rs1926447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:46055809 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTGTATACCTGGTATTTTTACTA[A/G]TTTTCTCAATAGCACGAACTGCTTC
Phenotype
MIM: 603101 MIM: 616453
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPB2 PubMed Links

Gene Details

Gene
CPB2
Gene Name
carboxypeptidase B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278541.1 1107 Missense Mutation ACT,ATT T310I NP_001265470.1
NM_001872.4 1107 Missense Mutation ACT,ATT T347I NP_001863.3
XM_017020393.1 1107 Missense Mutation ACT,ATT T338I XP_016875882.1
Gene
CPB2-AS1
Gene Name
CPB2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZC3H13
Gene Name
zinc finger CCCH-type containing 13
There are no transcripts associated with this gene.

View Full Product Details