Product Details
- SNP ID
-
rs13144
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:132769632 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGATCATAAGTGAACGTAGGTTAC[A/C]AAAATGGAAGTTGTAAGTATTGTTA
- Phenotype
-
MIM: 613361
MIM: 603571
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC18B1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80197282] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC18B1
- Gene Name
- solute carrier family 18 member B1
- Gene
- VNN2
- Gene Name
- vanin 2
There are no transcripts associated with this gene.
View Full Product Details