Product Details

SNP ID
rs4804572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11166398 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGGAGCTGGAGTCCTGTGGCCGT[C/T]GGGGCTCCCCCAGGGAAGCAGAGGG
Phenotype
MIM: 614610
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KANK2 PubMed Links

Gene Details

Gene
KANK2
Gene Name
KN motif and ankyrin repeat domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136191.2 3077 UTR 3 NP_001129663.1
NM_015493.6 3077 UTR 3 NP_056308.3

View Full Product Details