Product Details
- SNP ID
-
rs2058389
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:76003891 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGACCACACAGGTCATCAGGTGGAA[C/T]GCAGAATGCAGGCATCCCCATTGTG
- Phenotype
-
MIM: 616695
MIM: 616550
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
STYXL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76090562] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- STYXL1
- Gene Name
- serine/threonine/tyrosine interacting-like 1
- Gene
- TMEM120A
- Gene Name
- transmembrane protein 120A
There are no transcripts associated with this gene.
View Full Product Details