Product Details

SNP ID
rs1917940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:97008500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGATCAAAGAGAAATGTTACCA[A/G]AATTGTAACAGGTGAAGCCACAGTT
Phenotype
MIM: 600030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLX6 PubMed Links

Gene Details

Gene
DLX6
Gene Name
distal-less homeobox 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005222.3 Intron NP_005213.3
Gene
DLX6-AS1
Gene Name
DLX6 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details