Product Details

SNP ID
rs34187696
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:56222056 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CGGGTGACTGACTGGGCTCGCAGGG[A/C]CTGGGGAATGAATTGAATTCATCCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN5A PubMed Links

Gene Details

Gene
ZSCAN5A
Gene Name
zinc finger and SCAN domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322061.1 1359 Missense Mutation NP_001308990.1
NM_001322062.1 1359 Intron NP_001308991.1
NM_001322064.1 1359 Missense Mutation NP_001308993.1
NM_001322065.1 1359 Missense Mutation NP_001308994.1
NM_001322066.1 1359 Missense Mutation NP_001308995.1
NM_001322067.1 1359 Missense Mutation NP_001308996.1
NM_001322068.1 1359 Missense Mutation NP_001308997.1
NM_001322069.1 1359 Missense Mutation NP_001308998.1
NM_001322070.1 1359 Missense Mutation NP_001308999.1
NM_001322072.1 1359 Missense Mutation NP_001309001.1
NM_001322073.1 1359 Missense Mutation NP_001309002.1
NM_001322074.1 1359 Missense Mutation NP_001309003.1
NM_001322075.1 1359 Missense Mutation NP_001309004.1
NM_001322076.1 1359 Missense Mutation NP_001309005.1
NM_001322077.1 1359 Missense Mutation NP_001309006.1
NM_001322078.1 1359 Missense Mutation NP_001309007.1
NM_024303.2 1359 Intron NP_077279.1
XM_005259254.1 1359 Intron XP_005259311.1
XM_006723377.3 1359 Missense Mutation XP_006723440.1
XM_011527308.1 1359 Intron XP_011525610.1
XM_017027298.1 1359 Missense Mutation XP_016882787.1
XM_017027299.1 1359 Missense Mutation XP_016882788.1

View Full Product Details