Product Details

SNP ID
rs12609555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18008942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCTGCTTGTCTCTGTATCTTG[C/T]CCCCTGAAGTCCCGTCCCTCCACCC
Phenotype
MIM: 602982
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARRDC2 PubMed Links

Gene Details

Gene
ARRDC2
Gene Name
arrestin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025604.2 Intron NP_001020775.1
NM_001286826.1 Intron NP_001273755.1
NM_015683.1 Intron NP_056498.1
Gene
KCNN1
Gene Name
potassium calcium-activated channel subfamily N member 1
There are no transcripts associated with this gene.

View Full Product Details