Product Details
- SNP ID
-
rs10425463
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:20644064 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAGCCAGAGTCTGGCAGAGGTCCT[A/G]CTATTCCAGAGACCTGGAGGAAGGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF626
PubMed Links
Gene Details
- Gene
- ZNF626
- Gene Name
- zinc finger protein 626
View Full Product Details