Product Details

SNP ID
rs2040474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:18172799 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTACTGTTTAATACATATTTGAGA[A/C]ATTGACTTCAATTTAATTGAATTTC
Phenotype
MIM: 108345
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NAT1 PubMed Links

Gene Details

Gene
NAT1
Gene Name
N-acetyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000662.7 Intron NP_000653.3
NM_001160170.3 Intron NP_001153642.1
NM_001160171.3 Intron NP_001153643.1
NM_001160172.3 Intron NP_001153644.1
NM_001160173.3 Intron NP_001153645.1
NM_001160174.2 Intron NP_001153646.1
NM_001160175.3 Intron NP_001153647.1
NM_001160176.3 Intron NP_001153648.1
NM_001160179.2 Intron NP_001153651.1
NM_001291962.1 Intron NP_001278891.1
XM_006716410.3 Intron XP_006716473.1
XM_011544687.1 Intron XP_011542989.1
XM_011544688.1 Intron XP_011542990.1
XM_011544689.2 Intron XP_011542991.1
XM_017013947.1 Intron XP_016869436.1

View Full Product Details