Product Details

SNP ID
rs2609881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44327836 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCACTGAAACCCTTTGTACACACC[G/T]CACATTTGTATGGTTTCCCTCCTGT
Phenotype
MIM: 603994
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF112 PubMed Links

Gene Details

Gene
ZNF112
Gene Name
zinc finger protein 112
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083335.1 2452 Missense Mutation GAG,GCG E780A NP_001076804.1
NM_013380.3 2452 Missense Mutation GAG,GCG E774A NP_037512.3

View Full Product Details