Product Details

SNP ID
rs1924422
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:36966077 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TATTTTATGCTGCTTTGCCCCACAT[A/G]TATTTCAGTTTCAAAAAAGCTGTCA
Phenotype
MIM: 604565
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALG5 PubMed Links

Gene Details

Gene
ALG5
Gene Name
ALG5, dolichyl-phosphate beta-glucosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142364.1 Intron NP_001135836.1
NM_013338.4 Intron NP_037470.1
XM_017020552.1 Intron XP_016876041.1

View Full Product Details