Product Details

SNP ID
rs8107232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51345570 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTGGACAACTCCTGCTGGCTAGG[A/G]GGCCCTGGGAGGCCCAACTACTGCA
Phenotype
MIM: 130410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ETFB PubMed Links

Gene Details

Gene
ETFB
Gene Name
electron transfer flavoprotein beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014763.1 Intron NP_001014763.1
NM_001985.2 Intron NP_001976.1
Gene
VSIG10L
Gene Name
V-set and immunoglobulin domain containing 10 like
There are no transcripts associated with this gene.

View Full Product Details