Product Details
- SNP ID
-
rs1819191
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:49699834 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTTTGTGACCTTAATTAGCTTTTA[C/T]GTTGTTATTCCTACTTTTAGCGAAC
- Phenotype
-
MIM: 601892
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EBPL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141523087] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EBPL
- Gene Name
- emopamil binding protein like
There are no transcripts associated with this gene.
- Gene
- KPNA3
- Gene Name
- karyopherin subunit alpha 3
View Full Product Details