Product Details
- SNP ID
-
rs6509193
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:45097033 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTAGGCTCCATGCTGGGGAGGACG[A/G]TGGAGTGGTTCACTGGCCATGGGGC
- Phenotype
-
MIM: 607419
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GEMIN7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77027578] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GEMIN7
- Gene Name
- gem nuclear organelle associated protein 7
There are no transcripts associated with this gene.
- Gene
- LOC105372419
- Gene Name
- uncharacterized LOC105372419
There are no transcripts associated with this gene.
- Gene
- PPP1R37
- Gene Name
- protein phosphatase 1 regulatory subunit 37
View Full Product Details