Product Details

SNP ID
rs12610125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48997925 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGGAGAACTGTCAACATGGGAAG[A/C]CTCGGTATATTCTTGTTGAATGAGC
Phenotype
MIM: 138570 MIM: 604788
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GYS1 PubMed Links

Gene Details

Gene
GYS1
Gene Name
glycogen synthase 1
There are no transcripts associated with this gene.

Gene
RUVBL2
Gene Name
RuvB like AAA ATPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321190.1 Intron NP_001308119.1
NM_001321191.1 Intron NP_001308120.1
NM_006666.2 Intron NP_006657.1
XM_011526330.1 Intron XP_011524632.1

View Full Product Details