Product Details
- SNP ID
-
rs4802053
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39656599 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTAACTGGTAGGGATCTTGGAGAT[A/T]GTGAGTTACCAAGGACTTGTTTCAG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LGALS16
PubMed Links
Gene Details
- Gene
- LGALS16
- Gene Name
- galectin 16
View Full Product Details