Product Details

SNP ID
rs15540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:27547361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTACATGGTAAGACTTAGCAAGAA[A/G]AAAACAAGATGAAAATGACTGACCA
Phenotype
MIM: 614260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf72 PubMed Links

Gene Details

Gene
C9orf72
Gene Name
chromosome 9 open reading frame 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256054.2 2428 UTR 3 NP_001242983.1
NM_018325.4 2428 UTR 3 NP_060795.1
NM_145005.6 2428 Intron NP_659442.2

View Full Product Details