Product Details

SNP ID
rs183749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:49281603 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGCCGCTCTGGACACTACACCT[C/T]TTCCTCGCACTCCGGGACTAGCGTC
Phenotype
MIM: 600432
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CBLN1 PubMed Links

Gene Details

Gene
CBLN1
Gene Name
cerebellin 1 precursor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004352.3 229 UTR 5 NP_004343.1

View Full Product Details