Product Details

SNP ID
rs34929244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34685650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGAAGAGAAGCAGTGTTTATCA[C/T]GGTAAACTTCATTCATAGATCCTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCGB1B2P PubMed Links

Gene Details

Gene
SCGB1B2P
Gene Name
secretoglobin family 1B member 2, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF302
Gene Name
zinc finger protein 302
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012320.2 2031 UTR 3 NP_001012320.1
NM_001289181.1 2031 UTR 3 NP_001276110.1
NM_001289182.1 2031 UTR 3 NP_001276111.1
NM_001289183.1 2031 Intron NP_001276112.1
NM_001289184.1 2031 UTR 3 NP_001276113.1
NM_001289185.1 2031 UTR 3 NP_001276114.1
NM_001289186.1 2031 Intron NP_001276115.1
NM_001289187.1 2031 UTR 3 NP_001276116.1
NM_001289188.1 2031 UTR 3 NP_001276117.1
NM_001289189.1 2031 UTR 3 NP_001276118.1
NM_001289190.1 2031 Intron NP_001276119.1
NM_001289191.1 2031 UTR 3 NP_001276120.1
NM_001289192.1 2031 UTR 3 NP_001276121.1
NM_018443.3 2031 UTR 3 NP_060913.2
NM_018675.2 2031 UTR 3 NP_061145.2
XM_011527111.2 2031 UTR 3 XP_011525413.1
XM_017026978.1 2031 UTR 3 XP_016882467.1
XM_017026979.1 2031 UTR 3 XP_016882468.1
XM_017026980.1 2031 UTR 3 XP_016882469.1
XM_017026981.1 2031 Intron XP_016882470.1
XM_017026982.1 2031 Intron XP_016882471.1
XM_017026983.1 2031 Intron XP_016882472.1
XM_017026984.1 2031 Intron XP_016882473.1
XM_017026985.1 2031 Intron XP_016882474.1
XM_017026986.1 2031 UTR 3 XP_016882475.1

View Full Product Details