Product Details

SNP ID
rs11034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108162314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTTATTAAACATTACATTCAG[C/T]GTAAAGGCTTTAACCATCATAATCA
Phenotype
MIM: 606621
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFT57 PubMed Links

Gene Details

Gene
IFT57
Gene Name
intraflagellar transport 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018010.3 1701 UTR 3 NP_060480.1

View Full Product Details