Product Details

SNP ID
rs2284651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23412935 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGGTATTGGGCAGGGACAGGGCA[C/T]GGGAACAAAGGTGAGAGGGGTCTGA
Phenotype
MIM: 616096 MIM: 613613 MIM: 160710 MIM: 160760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MHRT PubMed Links

Gene Details

Gene
MHRT
Gene Name
myosin heavy chain-associated RNA transcript
There are no transcripts associated with this gene.

Gene
MIR208B
Gene Name
microRNA 208b
There are no transcripts associated with this gene.

Gene
MYH6
Gene Name
myosin heavy chain 6
There are no transcripts associated with this gene.

Gene
MYH7
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000257.3 Intron NP_000248.2
XM_017021340.1 Intron XP_016876829.1

View Full Product Details