Product Details

SNP ID
rs2056864
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:29196085 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GCAGTGAAATCACAAAGCAAACATT[T/G]TTTGGGGAACTAAGATAGTATCTAG
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00189 PubMed Links

Gene Details

Gene
LINC00189
Gene Name
long intergenic non-protein coding RNA 189
There are no transcripts associated with this gene.

View Full Product Details