Product Details

SNP ID
rs14973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132262884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGCATTCACTACAGATCCCTA[G/T]TTTTTACAACTGTCAACTGTACATT
Phenotype
MIM: 608465
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SETX PubMed Links

Gene Details

Gene
SETX
Gene Name
senataxin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015046.5 4327 UTR 3 NP_055861.3
XM_005272171.1 4327 UTR 3 XP_005272228.1
XM_005272172.2 4327 Intron XP_005272229.1
XM_005272173.2 4327 Intron XP_005272230.1
XM_011518404.2 4327 Intron XP_011516706.1
XM_011518405.2 4327 Intron XP_011516707.1
XM_011518406.2 4327 Intron XP_011516708.1
XM_011518407.1 4327 Intron XP_011516709.1
XM_011518408.2 4327 Intron XP_011516710.1
XM_017014495.1 4327 UTR 3 XP_016869984.1
XM_017014496.1 4327 UTR 3 XP_016869985.1
XM_017014497.1 4327 UTR 3 XP_016869986.1

View Full Product Details