Product Details

SNP ID
rs6714483
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:10305406 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTTTGAAAGTGCCTGTAATTACTGA[G/T]CTCTTGAGGTTTTATTGGATAATGT
Phenotype
MIM: 600207
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HPCAL1 PubMed Links

Gene Details

Gene
HPCAL1
Gene Name
hippocalcin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258357.1 Intron NP_001245286.1
NM_001258358.1 Intron NP_001245287.1
NM_001258359.1 Intron NP_001245288.1
NM_002149.3 Intron NP_002140.2
NM_134421.2 Intron NP_602293.1
XM_005246160.1 Intron XP_005246217.1
XM_005246161.1 Intron XP_005246218.1
XM_005246162.1 Intron XP_005246219.1
XM_005246163.1 Intron XP_005246220.1
XM_011510346.2 Intron XP_011508648.1
XM_011510347.1 Intron XP_011508649.1
XM_011510348.1 Intron XP_011508650.1
XM_011510349.1 Intron XP_011508651.1
XM_017003950.1 Intron XP_016859439.1
XM_017003951.1 Intron XP_016859440.1
XM_017003952.1 Intron XP_016859441.1
XM_017003953.1 Intron XP_016859442.1

View Full Product Details