Product Details

SNP ID
rs1991634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:130760736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGAGGACATGGTGAGGGGAACA[G/A]AGGGGAGGGACTCAGTCTGCCCTCT
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
AMER3 PubMed Links

Gene Details

Gene
AMER3
Gene Name
APC membrane recruitment protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105193.1 Intron NP_001098663.1
NM_001105194.1 Intron NP_001098664.1
NM_001105195.1 Intron NP_001098665.1
NM_152698.2 Intron NP_689911.2
Gene
POTEI
Gene Name
POTE ankyrin domain family member I
There are no transcripts associated with this gene.

View Full Product Details